Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cytochalasin B, Dihydro-

Catalog No. 2502251MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-022-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-022-51MG Supplier MilliporeSigma™ Supplier No. 2502251MG
Add to Cart
Add to Cart

Induces changes in cell morphology and motility with no effect on sugar transport.

Induces changes in cell morphology and motility with no effect on sugar transport. Used as specific marker in studies of high affinity cytochalasin binding sites unrelated to sugar transport. Disrupts the equilibrium between monomeric and polymeric actin.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins Changes in cell morphology and motility
CAS 39156-67-7
Molecular Formula C29H39NO5
Formula Weight 481.6g/mol
Solubility DMSO, EtOH, or MeOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.