Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set IV

Catalog No. 5391361SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-913-61SET 1 Set
53-913-61ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-913-61SET Supplier MilliporeSigma™ Supplier No. 5391361SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Protease Inhibitor Cocktail Set IV controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

A cocktail of four protease inhibitors with broad specificity for the inhibition of aspartic-, cysteine-, metallo-, and serine-proteases. Recommended for fungal and yeast cell extracts. Each vial contains 100mm AEBSF, HCl (Cat. No. 101500), 1.5mm E-64 (Cat. No. 324890), 2mm Pepstatin A (Cat. No. 516481), and 500mm 1,10-Phenanthroline (Cat. No. 516705). Supplied with a data sheet. Note: 1 set = 5 x 1 ml.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 1mL DMSO.
Target Proteins aspartic-, cysteine-, metallo-, and serine-proteases
Form Liquid
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.