Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set V, EDTA-Free

Catalog No. 5391371ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
10 Vials
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-913-71ML 1 mL
53-913-710VL 10 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-913-71ML Supplier MilliporeSigma™ Supplier No. 5391371ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases.

A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases. Reconstitute each vial with 1mL H2O to obtain a 100X stock solution. 1X stock solution contains 500μM AEBSF, HCl (Cat. No. 101500), 150nM Aprotinin (Cat. No. 616370), 1μM E-64 (Cat. No. 324890), and 1μM Leupeptin Hemisulfate (Cat. No. 108975). Supplied with a data sheet.
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins Serine and cysteine proteases, but not metalloproteases
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.