Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ D-Amino Acid Oxidase Inhibitor III, AS057278

Catalog No. 25140650MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-140-650MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-140-650MG Supplier MilliporeSigma™ Supplier No. 25140650MG
Add to Cart
Add to Cart

A cell-permeable pyrazolocarboxylic acid that acts as a selective D-amino acid oxidase (DAO/DAAO/DAMOX/OXDA) inhibitor (IC50 = 910nM against human DAO; [D-ser] = 10 mM) and effectively protects DAO overexpressing cultures from oxidative stress-induced cell death upon D-Ser (50 mM) exposure (ED50

A cell-permeable pyrazolocarboxylic acid that acts as a selective D-amino acid oxidase (DAO/DAAO/DAMOX/OXDA) inhibitor (IC50 = 910nM against human DAO; [D-ser] = 10 mM) and effectively protects DAO overexpressing cultures from oxidative stress-induced cell death upon D-Ser (50 mM) exposure (ED50 ≤3.95μM), while exhibiting little affinity toward NMDAR glycine binding site or inhibitory activity against D-aspartate oxidase (DDO/DSOX). Reported to be orally available and blood-brain barrier-permeant in rats and effectively prevent PCP (phencyclidine) from suppressing PPI (prepulse inhibition) response in mice (single 80mg/kg or 56X 20mg/kg/12 h b.i.d. oral dosages) in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins DAAO
CAS 402-61-9
Molecular Formula C5H6N2O2
Formula Weight 126.11g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.