Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ D,L-Sulforaphane

Catalog No. 57421525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-421-525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-421-525MG Supplier MilliporeSigma™ Supplier No. 57421525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An isothiocyanate isolated from broccoli that acts as a potent inducer of phase II detoxifying enzymes in mouse tissues and murine hepatoma cells in culture.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Slight Yellow
Description 98% by UPLC
Target Proteins Potent inducer of phase II detoxifying enzymes in mouse tissues and murine hepatoma cells in culture
CAS 4478-93-7
Molecular Formula C6H11NOS2
Formula Weight 177.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Liquid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.