Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ D-Val-Phe-Lys Chloromethyl Ketone, Dihydrochloride

Catalog No. 6276245MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-762-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 62-762-45MG Supplier MilliporeSigma™ Supplier No. 6276245MG
Add to Cart
Add to Cart

Selective irreversible inhibitor of plasmin with high selectivity for plasmin (IC50 = 100 pM for human plasmin) over urokinase.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-D-Val-Phe-Lys-CMK
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 90% by TLC
Target Proteins Human plasmin
Formula Weight 497.9g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.