Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ D609, Potassium Salt

Catalog No. 2514005MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-140-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-140-05MG Supplier MilliporeSigma™ Supplier No. 2514005MG
Add to Cart
Add to Cart

Selective inhibitor of phosphatidylcholine (PC)-specific phospholipase C (Ki = 5-10 μM).

Selective inhibitor of phosphatidylcholine (PC)-specific phospholipase C (Ki = 5-10μM). Does not inhibit phosphatidylinositol-specific phospholipase C, phospholipase A, or phospholipase D. Also reported to inhibit LPS-stimulated ERK activation via its inhibitory effect on PLC. Inhibits angiogenesis by preventing the synthesis of basement membrane.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins pc-specificphopholipase c
CAS 83373-60-8
Molecular Formula C11H15OS2·K
Formula Weight 266.5g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.