Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DAB, Tetrahydrochloride, 50X Concentrate

Catalog No. 28175110ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-175-110ML 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-175-110ML Supplier MilliporeSigma™ Supplier No. 28175110ML
Add to Cart
Add to Cart

Used in Immunoblotting, Immunohistochemistry applications. MilliporeSigma™; Calbiochem™ DAB, Tetrahydrochloride, 50X Concentrate produces a brown alcohol-insoluble end product.

  • Mucin exists as an aggregated complex in excess of 106 molecular weight
  • Rich source of O-linked oligosaccharide chains
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
For Use With (Application) Immunoblotting, Immunohistochemistry
Quantity 10mL
Format Dark brown solution. Supplied as a 50X concentrate.
Product Type DAB, Tetrahydrochloride, 50X Concentrate
Storage Requirements Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.