Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Dephostatin

Catalog No. 2632001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-320-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-320-01MG Supplier MilliporeSigma™ Supplier No. 2632001MG

Protein tyrosine phosphatase inhibitor.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Description 90% by HPLC
Target Proteins protein tyrosine phosphatase activity from human neoplastic cell line
CAS 151606-30-3
Molecular Formula C7H8N2O3
Formula Weight 168.2g/mol
Solubility Chloroform, DMSO, or MeOH
Content And Storage Protect from light
Form Powder
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.