Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Dermatan Sulfate, Porcine Intestinal Mucosa

Catalog No. 263301250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-330-1250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-330-1250MG Supplier MilliporeSigma™ Supplier No. 263301250MG

Mucopolysaccharide containing iduronic acid, known to inhibit adenylate kinase from rat hepatic membranes and acts as potent mediator of fibroblast growth factor-2 responsiveness

  • Iduronic acid-containing mucopolysaccharide abundant in skin, arterial walls, and heart valves
  • Shown to inhibit both Gpp(NH)p- and FSK-stimulated adenylate cyclase activity in rat hepatic membrane preparations
  • Acts as an active mediator of fibroblast growth factor-2 responsiveness in wound healing via FGF-2 affinity interaction (KD 2.48 μM)
  • M.W. 30,000 (avg.)
TRUSTED_SUSTAINABILITY

Specifications

CAS 54328-33-5
Color White
Content And Storage Store at +2°C to +8°C
Quantity 250 mg
Synonym Iduronic Acid-β-1,4-N-acetyl-D-galactosamine, Chrondroitin Sulfate B
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.