Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Diacylglycerol Kinase Inhibitor I

Catalog No. 2667855MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-678-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-678-55MG Supplier MilliporeSigma™ Supplier No. 2667855MG
Add to Cart
Add to Cart

The Diacylglycerol Kinase Inhibitor I, also referenced under CAS 93076-89-2, controls the biological activity of Diacylglycerol Kinase. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.

Potentiates the activity of protein kinase C by inhibiting diacylglycerol metabolism. Inhibits diacylglycerol kinase (IC50 = 2.8μM) without affecting phosphodiesterase or phosphatidylinositol kinase. Weak dopamine D2 and histamine H1 antagonist and strong serotonin S2 antagonist.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Diacylglycerol Kinase
CAS 93076-89-2
Molecular Formula C27H26FN3OS
Formula Weight 459.6g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.