Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Diacylglycerol Kinase Inhibitor II

Catalog No. 2667885MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-678-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-678-85MG Supplier MilliporeSigma™ Supplier No. 2667885MG
Add to Cart
Add to Cart

The Diacylglycerol Kinase Inhibitor II, also referenced under CAS 120166-69-0, controls the biological activity of Diacylglycerol KInase. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.

Inhibits diacylglycerol kinase in isolated platelet membranes (IC50 = 300 nM) and in intact platelets (IC50 = 120 nM) by binding to the catalytic domain. A more potent analog of diacylglycerol kinase inhibitor I (Cat. No. 266785).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Diacylglycerol Kinase
CAS 120166-69-0
Molecular Formula C28H25F2N3OS
Formula Weight 489.6g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.