Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DIDS, Disodium Salt

Catalog No. 309795250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-979-5250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30-979-5250MG Supplier MilliporeSigma™ Supplier No. 309795250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Binds covalently and irreversibly to the outer surface of human erythrocyte membranes.

Binds covalently and irreversibly to the outer surface of human erythrocyte membranes. Inhibits ATP transport into endoplasmic reticulum vesicles. Anion transport inhibitor that inhibits Cl uptake (IC50 = 150 nM) in neuroblastoma cells. Has anti-ulcerous properties.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 85% by TLC
Target Proteins Cl- uptake
CAS 53005-05-3
Molecular Formula C16H8N2O6S4·2Na
Formula Weight 498.5g/mol
Solubility H2O
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.