Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Dipeptidylpeptidase IV Inhibitor I

Catalog No. 4162005MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-620-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-620-05MG Supplier MilliporeSigma™ Supplier No. 4162005MG
Add to Cart
Add to Cart

The Dipeptidylpeptidase IV Inhibitor I controls the biological activity of Dipeptidylpeptidase IV. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Peptide Sequence: Ile-Pro-Ile
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins dipeptidylpeptidase IV (DPP IV)
Molecular Formula C17H31N3O4
Formula Weight 341.5g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic
Form Amorphous Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.