Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Dipeptidylpeptidase IV Inhibitor IV, K579

Catalog No. 3176425MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-764-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-764-25MG Supplier MilliporeSigma™ Supplier No. 3176425MG
Add to Cart
Add to Cart

The Dipeptidylpeptidase IV Inhibitor IV, K579, also referenced under CAS 440100-64-1, controls the biological activity of Dipeptidylpeptidase IV. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Light Brown
Description 97% by HPLC
CAS 440100-64-1
Molecular Formula C17H24N6O
Formula Weight 328.4g/mol
Solubility DMSO, EtOH, or 1 eq HCl
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.