Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Diphenyleneiodonium Chloride

Catalog No. 30026010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-026-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30-026-010MG Supplier MilliporeSigma™ Supplier No. 30026010MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable, irreversible inhibitor of endothelial nitric oxide synthase (eNOS).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
Target Proteins eNOS
CAS 4673-26-1
Molecular Formula C12H8ICl
Formula Weight 314.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Crystalline Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.