Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Dkk Inhibitor II, IIIC3a

Catalog No. 31770125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-770-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-770-125MG Supplier MilliporeSigma™ Supplier No. 31770125MG
Add to Cart
Add to Cart

An enhanced in solution stable gallocyanine analog that directly and reversibly targets human LRP5/6 (low density lipoprotein-related protein 5/6), and disrupts its interaction with Dkk (Dickkopf) in a competitive manner

An enhanced in solution stable gallocyanine analog that directly and reversibly targets human LRP5/6 (low density lipoprotein-related protein 5/6), and disrupts its interaction with Dkk (Dickkopf) in a competitive manner. Shown to potently reverse DKK1/2/4-mediated inhibition of WNT3A-stimulated reporter gene activity (IC50 ∽ 10 - 30μM in in NIH 3T3 cells) with no effects on sFRP4-mediated inhibition of Wnt signaling. Blocks Dkk-2 - alkaline phosphatase binding to cells expressing LRP5 (IC50 = 1μM in LRP5 transfected NIH 3T3 cells). Reduces blood glucose levels, improves glucose tolerance, and increases insulin sensitivity in type 2 diabetic mice (1.5mg/kg, i.p. daily), mainly via its antagonistic action on DKK2. These effects are not observed in DKK2 -/- mice.
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Description 90% by HPLC
Target Proteins LRP5/6
Molecular Formula C16H16I2N2O5
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.