Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DL-α-Tocopherol

Catalog No. 61342050GM Shop All MilliporeSigma Products
Change view
Click to view available options
DL-α-Tocopherol; Qty: 50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
61-342-050GM DL-α-Tocopherol; Qty: 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 61-342-050GM Supplier MilliporeSigma™ Supplier No. 61342050GM

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protects cell membrane lipids from oxidative damage by free radicals

  • Physiological membrane-bound antioxidant
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Additive DL-α-Tocopherol
Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.