Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DNA Base Excision Repair Pathway Inhibitor

Catalog No. 26201525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-201-525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-201-525MG Supplier MilliporeSigma™ Supplier No. 26201525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DNA Base Excision Repair Pathway Inhibitor, also referenced under CAS 6960-45-8, controls the biological activity of APE1 (human apurinic/apyrimidinic endonuclease.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 95% by titration
Target Proteins APE 1
CAS 6960-45-8
Molecular Formula C9H6N2O4
Formula Weight 206.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.