Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DNA Methyltransferase Inhibitor

Catalog No. 26092010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-092-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-092-010MG Supplier MilliporeSigma™ Supplier No. 26092010MG
Add to Cart
Add to Cart

The DNA Methyltransferase Inhibitor, also referenced under CAS 32675-71-1, controls the biological activity of DNA Methyltransferase. This small molecule/inhibitor is primarily used for Cell Structure applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins CPG methylase M.Sssl
CAS 32675-71-1
Molecular Formula C19H14N2O4
Formula Weight 334.3g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.