Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DNQX Disodium Salt

Catalog No. 5050260001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-502-60001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-502-60001 Supplier MilliporeSigma™ Supplier No. 5050260001
Add to Cart
Add to Cart

A potent competitive antagonist selective for AMPA/kainate glutamate receptors (IC50

A potent competitive antagonist selective for AMPA/kainate glutamate receptors (IC50 = 0.19-1.66μM and 0.35μM for AMPA and GluK2 receptors, respectively). Routinely used as a tool to distinguish the role of neurotransmissions mediated by AMPA/kainate and NMDA receptors in studying brain functions and disorders.
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Brown
Description 99% by HPLC
Target Proteins AMPA
Molecular Formula C8H2N4O6Na2
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.