Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DPDPE

Catalog No. 5081600001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-816-00001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-816-00001 Supplier MilliporeSigma™ Supplier No. 5081600001
Add to Cart
Add to Cart

A potent agonist selective for δ opioid receptor (EC50

A potent agonist selective for δ opioid receptor (EC50 = 5.2nM). Exhibits anti-convulsant effects in vivo in rats, also used in nociceptive studies.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins δ opioid receptor
Molecular Formula C30H39N5O7S2
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.