Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DUB Inhibitor XI

Catalog No. 66214310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-214-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-214-310MG Supplier MilliporeSigma™ Supplier No. 66214310MG
Add to Cart
Add to Cart

A cell-permeable compound that acts as a dual inhibitor of USP7 (IC50 = 0.42 μM) and USP47 (IC50 = 1.0 μM).

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 95% by HPLC
Target Proteins USP7/47
Molecular Formula C18H11Cl2N3O3S3
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.