Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Dynamin Inhibitor III, Phthaladyn-23

Catalog No. 32441210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-441-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-441-210MG Supplier MilliporeSigma™ Supplier No. 32441210MG
Add to Cart
Add to Cart

A cell-permeable phthalimide compound that is reported to inhibit dynamin GTPase activity (IC50 =

A cell-permeable phthalimide compound that is reported to inhibit dynamin GTPase activity (IC50 = 17.4 and 63μM using purified sheep brain dynamin I and recombinant dynamin II, respectively) in a GTP-competitive manner. Shown to block K+-induced SVE (synaptic vesicle endocytosis) of FM4-64 dye into rat brain synaptosomes (IC50 = 12.9μM), while being ineffective at 30μM against RME (receptor-mediated endocytosis) of transferrin into human osteosarcoma epithelial U2OS cells, presumably due to poor dynamin II inhibitory potency and/or rapid efflux and/or limited permeability in this cell line.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow to Beige
Description 97% by HPLC
Molecular Formula C22H12ClN3O7
Formula Weight 465.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.