Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Dynamin Inhibitor IV, Hydroxy-Dynasore

Catalog No. 32441310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-441-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-441-310MG Supplier MilliporeSigma™ Supplier No. 32441310MG
Add to Cart
Add to Cart

The Dynamin Inhibitor IV, Hydroxy-Dynasore controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by HPLC
Molecular Formula C18H14N2O5
Formula Weight 338.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.