Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ E-4031

Catalog No. 3244705MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-447-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-447-05MG Supplier MilliporeSigma™ Supplier No. 3244705MG
Add to Cart
Add to Cart

A potent and selective inhibitor of the rapid type of inactivating inward-rectifying K+ current (IK, IR) (IC50 = 10 nM).

A potent and selective inhibitor of the rapid type of inactivating inward-rectifying K+ current (IK, IR) (IC50 = 10 nM). A class III anti-arrhythmic agent with differential atrial versus ventricular activity. A specific blocker of HERG channels (IC50 = 7.7 nM).
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by TLC
Target Proteins Inhibitor of the rapid type of inactivating inward-rectifying K+ current
CAS 113558-89-7
Molecular Formula C21H27N3O3S·2HCl·2H2O
Formula Weight 510.5g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.