Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ E-Selectin, Human, Recombinant, CHO Cells

Catalog No. 56130050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-130-050UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-130-050UG Supplier MilliporeSigma™ Supplier No. 56130050UG
Add to Cart
Add to Cart

Transiently expressed on vascular endothelial cell surfaces

  • Recombinant
  • Consisting of 535 amino acids minus the transmembrane and cytoplasmic domains expressed in CHO cells
  • Initial binding event is followed by second interaction involving ICAM and VCAM-1 that leads to vascular penetration and eukocyte invasion into extracellular matrix tissue
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 μg
Product Type E-Selectin, Human, Recombinant, CHO Cells
Content And Storage -20°C
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.