Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Eeyarestatin I

Catalog No. 32452125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-452-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-452-125MG Supplier MilliporeSigma™ Supplier No. 32452125MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable oxo-imidazolidinyl-hydroxyurea that localizes to ER, where it interacts with AAA ATPase p97 via its nitrofuran-containing moiety, without exhibiting affinity toward Hsp70 / ATPase NSF

A cell-permeable oxo-imidazolidinyl-hydroxyurea that preferentially localizes to ER, where it interacts with AAA (ATPase associated with diverse cellular activities) ATPase p97 ( K d = 5 - 10μM) via its nitrofuran-containing moiety, without exhibiting affinity toward Hsp70 or AAA ATPase NSF (N-methylmaleimide-sensitive factor). Evidence indicates that EerI cellular metabolite, but not EerI itself, is responsible for the inhibition of ER membrane translocon sec61 complex-mediated transfer of newly synthesized polypeptide, resulting in a blockage of ER-mediated posttranslational glycosylation and signal peptide removal (Effective conc. = 8μM in HepG2 and HeLa cultures). Ether independent or as a consequence of the early effect on sec61 complex, EerI culture treatment also induces a selective dissociation of an 180 kDa protein from the atx3-containing p97/VCP (Valosin-containing protein) complex and a blockage of the complex-associated deubiquitination of ERAD (ER-associated protein degradation) substrates in a reversible manner, resulting in an accumulation of polyubiquitinated proteins (Effective conc. = 10μM in A9 and 293T cultures). EerI culture treatment (10μM) is also demonstrated to selectively induce cytotoxicity in lymphoid cell lines, BJAB, HBL-2, JEKO-1, Jurkat, KMS-12, MINO, as well as primary leukemia cells from CLL (chronic lymphocytic leukemia) patients, but not PBMC from healthy donors, by upregulating the BH3-only pro-apoptotic protein NOXA in cancer cells via ATF3/4 activation and a downregulation of H2A ubiquitination. Unlike DBeQ (Cat. No. 506190), EeRI does not inhibit p97 ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow-orange
Description 90% by HPLC
CAS 412960-54-4
Molecular Formula C27H25Cl2N7O7
Formula Weight 630.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.