Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Eg5 Inhibitor IV, VS-83

Catalog No. 3246235MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-462-35MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-462-35MG Supplier MilliporeSigma™ Supplier No. 3246235MG
Add to Cart
Add to Cart

The Eg5 Inhibitor IV, VS-83, also referenced under CAS 909250-29-9, controls the biological activity of Eg5. This small molecule/inhibitor is primarily used for Cell Structure applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
CAS 909250-29-9
Molecular Formula C14H11FN2OS
Formula Weight 274.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.