Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ EGFR Inhibitor II, BIBX1382

Catalog No. 3248325MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-483-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-483-25MG Supplier MilliporeSigma™ Supplier No. 3248325MG

The EGFR Inhibitor II, BIBX1382, also referenced under CAS 196612-93-8, controls the biological activity of EGFR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by HPLC
Target Proteins EGFR (ErbB-1, HER-1)
CAS 196612-93-8
Molecular Formula C18H19ClFN7·2HCl·2H2O
Formula Weight 496.8g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.