Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ EIF2AK3 Activator, CCT020312

Catalog No. 3248795MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-487-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-487-95MG Supplier MilliporeSigma™ Supplier No. 3248795MG
Add to Cart
Add to Cart

The EIF2AK3 Activator, CCT020312 modulates the biological activity of EIF2AK3.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish White
Description 98% by HPLC
Molecular Formula C31H30Br2N4O2
Formula Weight 650.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.