Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Elastase Inhibitor II

Catalog No. 3247445MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-474-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-474-45MG Supplier MilliporeSigma™ Supplier No. 3247445MG
Add to Cart
Add to Cart

The Elastase Inhibitor II controls the biological activity of Elastase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Peptide Sequence: MeOSuc-Ala-Ala-Pro-Ala-CMK
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by TLC
Target Proteins Human neutrophil elastase (HNE)
Molecular Formula C20H31ClN4O7
Formula Weight 474.9g/mol
Solubility DMSO or MeOH
Content And Storage Protect from moisture
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.