Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Elastase Inhibitor V

Catalog No. 32476125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-476-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-476-125MG Supplier MilliporeSigma™ Supplier No. 32476125MG
Add to Cart
Add to Cart

A benzoxazinone compound that acts as a potent inhibitor of human leukocyte elastase (IC50 =

A benzoxazinone compound that acts as a potent inhibitor of human leukocyte elastase (IC50 = 29.5nM), likely by covalently modifying active site serine. Shown to inhibit elastase secretion (IC50 = 80.8nM), but not O2 generation, in fLMP -stimulated primary human neutrophil cultures. Reported to reduce neutrophil infiltration in lung after hemorrhagic shock in rats in vivo (1mg/kg, i.v.).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins Elastase
Molecular Formula C14H7BrClNO2
Formula Weight 336.6g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.