Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Elastase Substrate V, Fluorogenic

Catalog No. 32474025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-474-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-474-025MG Supplier MilliporeSigma™ Supplier No. 32474025MG
Add to Cart
Add to Cart

Elastase Substrate V, Fluorogenic, CAS 72252-90-5, is a sensitive fluorogenic substrate for the quantitative determination of human leukocyte and porcine pancreatic elastase (Km = 140 μM).

Peptide Sequence: MeOSuc-Ala-Ala-Pro-Val-AMC
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Sensitive fluorogenic substrate for the quantitative determination of human leukocyte and porcine pancreatic elastase
Molecular Formula C31H41N5O9
Formula Weight 627.7g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Lyophilized
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.