Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Endothall

Catalog No. 32476020MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-476-020MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-476-020MG Supplier MilliporeSigma™ Supplier No. 32476020MG

A cell-permeable, specific inhibitor of protein phosphatase 2A (PP2A, IC50 = 90 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins PP2A
CAS 145-73-3
Molecular Formula C8H10O5
Formula Weight 186.2g/mol
Solubility H2O
Form Solid
Quantity 20 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.