Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Endothelial Cell Growth Supplement, Bovine Neural Tissue

Catalog No. 32484515MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
15 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-484-515MG 15 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-484-515MG Supplier MilliporeSigma™ Supplier No. 32484515MG
Only null left
Add to Cart
Add to Cart

Used in screening for contamination by bacteria, fungi, and mycoplasm

  • Acts as superior cloning factor for isolation and growth of hybridoma cells
  • Eliminates need for feeder cell layers
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered NaCl containing streptomycin sulfate.
Quantity 15 mg
Storage Requirements 2° to 8°C
Product Type Endothelial Cell Growth Supplement, Bovine Neural Tissue
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.