Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ERK Activation Inhibitor Peptide I, Cell-Permeable

Catalog No. 3280001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-800-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-800-01MG Supplier MilliporeSigma™ Supplier No. 3280001MG
Add to Cart
Add to Cart

The ERK Activation Inhibitor Peptide I, Cell-Permeable controls the biological activity of ERK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ste-Met-Pro-Lys-Lys-Lys-Pro-Thr-Pro-Ile-Gln-Leu-Asn-Pro-NH2 (Ste = stearated)
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins MEK
Molecular Formula C86H153N19O17S
Formula Weight 1757.3g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.