Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ERK Inhibitor VIII

Catalog No. 32801110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-801-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-801-110MG Supplier MilliporeSigma™ Supplier No. 32801110MG
Add to Cart
Add to Cart

A cell-permeable pyrimidylpyrrole compound that acts as an active site-targeting, highly potent and selective ERK1/2 inhibitor ( K I <2nM against Erk2; [ATP] = 65μM), while inhibiting GSK-3, Aurora A, Cdk2 only at much higher concentrations ( K I = 395, 540, and 852 nM, respectively) and exhibiting much reduced or little potency toward a panel of more than 130 other kinases ( K

A cell-permeable pyrimidylpyrrole compound that acts as an active site-targeting, highly potent and selective ERK1/2 inhibitor ( K I <2nM against Erk2; [ATP] = 65μM), while inhibiting GSK-3, Aurora A, Cdk2 only at much higher concentrations ( K I = 395, 540, and 852 nM, respectively) and exhibiting much reduced or little potency toward a panel of more than 130 other kinases ( K I ≥1.4μM; IC50 ≥1μM). Shown to effectively inhibit human colon carcinoma HT-29 proliferation (IC50 = 48nM) and restore EGFR inhibitor WZ4002 antiproliferation activity in WZR10 cultures (84% and no inhibition, respectively, by 100nM WZ4002 with or without 1μM Erk Inhibitor VIII co-treatment). Orally available in both mice and rats in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish-white
Description 98% by Chiral HPLC
Target Proteins ERK
Molecular Formula C24H20Cl2FN5O2
Formula Weight 500.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.