Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ERO1 Inhibitor II, EN460

Catalog No. 32850125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-850-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-850-125MG Supplier MilliporeSigma™ Supplier No. 32850125MG
Add to Cart
Add to Cart

A cell-permeable thiol reactive enone (EN) compound that selectively interacts with the active-site cysteine of reduced, active form of ERO1α and inhibits its activity (IC50

A cell-permeable thiol reactive enone (EN) compound that selectively interacts with the active-site cysteine of reduced, active form of ERO1α and inhibits its activity (IC50 = 1.9μM). Also prevents ERO1 re-oxidation both in vitro and in mouse embryonic fibroblasts. Activates the unfolded protein response and protects ER-stressed 293T cells. Can inhibit ERO1α even in the presence of an excess amount of competing thiols. Can inhibit ERO1α even in the presence of an excess amount of competing thiols. EN460 binding to ERO1α is shown to promote the loss of flavin adenine dinucleotide (FAD) from the holoenzyme. Its inhibitory action appears to be irreversible, however addition of FAD and tris(hydroxypropyl)phosphine (Cat. No. 598250) can restore some enzyme activity.
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description 99% by HPLC
Molecular Formula C22H12ClF3N2O4
Formula Weight 460.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.