Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Etomoxir

Catalog No. 2360205MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-602-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-602-05MG Supplier MilliporeSigma™ Supplier No. 2360205MG
Add to Cart
Add to Cart

A cell-permeable, irreversible, and stereospecific compound that is shown to inhibit carnitine palmitoyltransferase (CPT)-1 and DGAT activity in the mitochondria of rat heart H9c2 myoblastic cells.

A cell-permeable, irreversible, and stereospecific compound that is shown to inhibit carnitine palmitoyltransferase (CPT)-1 and DGAT activity in the mitochondria of rat heart H9c2 myoblastic cells at a concentration of 1-80μM and 40μM, respectively. Also available as a 50mm solution in H2O (Cat. No. 509455).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
CAS 828934-41-4
Molecular Formula C15H18ClO4·Na·H2O
Formula Weight 338.76g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.