Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Etoposide

Catalog No. 34120525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-120-525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-120-525MG Supplier MilliporeSigma™ Supplier No. 34120525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etoposide, CAS 33419-42-0, is cell-permeable inhibitor of topoisomerase II inhibitor (IC50 = 59.2μM). Displays anti-tumor activity.

A cell-permeable derivative of podophyllotoxin that acts as a topoisomerase II inhibitor (IC50 = 59.2μM) has major activity against a number of tumors, including germ cell neoplasms, small cell lung cancer, and malignant lymphoma. Induces apoptosis in mouse thymocytes and in HL-60 human leukemia cells.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description >97% by HPLC
Product Type Inhibitor
Target Proteins topoisomerase 2
CAS 33419-42-0
Molecular Formula C29H32O13
Formula Weight 588.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.