Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Eya2 Phosphatase Inhibitor, MLS000544460

Catalog No. 5310850001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-108-50001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-108-50001 Supplier MilliporeSigma™ Supplier No. 5310850001
Add to Cart
Add to Cart

A cell-permeable furylidenebenzohydrazide compound that acts as an inhibitor of Eya2 ED tyrosine phosphatase (IC50

A cell-permeable furylidenebenzohydrazide compound that acts as an inhibitor of Eya2 ED tyrosine phosphatase (IC50 = 4.1μM; KD = 2.0μM for hEya2, catalytic domain at 5μM Mg2+). Inhibition appears to be reversible and allosteric in nature. Does not affect the activity of Eya3, PTP1B, PPM1A, and Scp1 even at higher concentration (IC50 > 100μM). Although its binding to Eya2 ED is not Mg2+ dependent, its inhibitory activity is dependent on the concentration of Mg2+ in the medium. At higher Mg2+ levels (∽5 mM) the inhibition is non-competitive whereas at low Mg2+ levels (∽5μM) it has a mixed type inhibition. Reduces Eya2-mediated cell migration in MCF10A cells (∽ 10μM).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Description 97% by HPLC
Target Proteins Eya2 C-terminal Eya domain (ED) protein tyrosine phosphatase
Molecular Formula C17H12FN3O2S
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.