Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ezh2/Ezh1 Inhibitor, UNC1999

Catalog No. 5050520001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-505-20001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-505-20001 Supplier MilliporeSigma™ Supplier No. 5050520001
Add to Cart
Add to Cart

A cell-permeable GSK126 structural analog

A cell-permeable GSK126 structural analog that acts as a potent, SAM-, but not substrate-, competitive inhibitor against Ezh1 and Ezh2 (IC50 = 5 and 45nM against PRC2 complex containing Ezh2 or Ezh1 in H3 peptide methylation assays; [SAM] = 5μM), while exhibiting much reduced or little potency toward panels of 15 other methyltransferases, 50 kinases, as well as 43 GPCRs, transporters, and ion channels, although high affinity binding is observed with Sigma-2 receptor S2R/PGRMC1 ( K i = 65nM) with unknown impact toward its function. Cellular H3K27me3 level is shown to be largely reduced in MCF10A, MCF7, and DB cells with no significant effect on proliferation upon a 3-day UNC1999 treatment (ICmax = 3 to 5μM), while complete cell death is reported in the EZH2 Y641N-harboring DB cell line upon an 8-day 5μM drug exposure. Both its oral bioavailability in mice and enhanced Ezh1 potency can greatly complement the Ezh2-selective GSK126 in in vitro and in vivo studies.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Ezh2
Molecular Formula C33H43N7O2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.