Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ezh2 Inhibitor III, GSK126

Catalog No. 5005800001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-058-00001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-058-00001 Supplier MilliporeSigma™ Supplier No. 5005800001
Add to Cart
Add to Cart

A cell-permeable structural analog of E1 that acts as an equipotent, SAM-competitive inhibitor of Ezh2/PRC2 inhibitor (Ki = 0.5 to 3 nM against wt & mutant Ezh2-containing PRC2).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Molecular Formula C31H38N6O2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.