Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ezh2 Inhibitor III, SAH-EZH2

Catalog No. 5083200001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-832-00001 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-832-00001 Supplier MilliporeSigma™ Supplier No. 5083200001
Add to Cart
Add to Cart

A helical 27-mer peptide with E54Q modification that directly binds to EDD (Kd = 320 nM) and disrupts protein-protein interaction of EED with EZH2 and EZH1.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-FSSNRXKILXRTQILNQEWKQRRIQPV (X=(S)-2-(4′-pentenyl)Ala, stapled)
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins EED (embryonic ectoderm development) - EZH2
Molecular Formula C155H256N48O40·6TFA·14H2O
Solubility H2O
Content And Storage Protect from light
Form Powder
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.