Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ FATP2 Inhibitor, CB-2

Catalog No. 5006700001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-067-00001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-067-00001 Supplier MilliporeSigma™ Supplier No. 5006700001
Add to Cart
Add to Cart

A cell permeable thioxothiazolindine compound that reversibly inhibits FATP2-mediated long chain fatty acid uptake in Caco-2 and HepG2, but not non-FATP2-expressing 3T3-L1 adipocytes, cultures (IC50

A cell permeable thioxothiazolindine compound that reversibly inhibits FATP2-mediated long chain fatty acid uptake in Caco-2 and HepG2, but not non-FATP2-expressing 3T3-L1 adipocytes, cultures (IC50 = 3.99, 7.55, and 231.1μM, respectively), while exhibiting much reduced potency toward cellular Acsl (acyl-CoA synthetase) activity (IC50 = 50μM). Shown not to affect Caco-2 viability, membrane permeability, or glucose transport.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 99% by HPLC
Molecular Formula C17H11BrClNO3S2
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.