Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Fbxo3 Inhibitor, BC-1215

Catalog No. 5048430001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-484-30001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-484-30001 Supplier MilliporeSigma™ Supplier No. 5048430001
Add to Cart
Add to Cart

A cell-permeable, bis-pyridinylbenzylethanamine that disrupts Fbxo3-Fbxl2 interaction and effectively prevents SCF-Fbxo3-catalyzed Fbxl2 ubiquitination, resulting in cellular Fbxl2 upregulation and thereby SCF-Fbxl2-catalyzed TRAFs (TNF Receptor-Associated Factors) ubiquitination

A cell-permeable, bis-pyridinylbenzylethanamine that disrupts Fbxo3-Fbxl2 interaction and effectively prevents SCF-Fbxo3-catalyzed Fbxl2 ubiquitination, resulting in cellular Fbxl2 upregulation and thereby SCF-Fbxl2-catalyzed TRAFs (TNF Receptor-Associated Factors) ubiquitination. Effectively reduces cellular TRAFs (5 to 127μM) and prevents TRAF-mediated cytokines production from LPS-stimulated human PBMC (25μM). Shown to greatly prevent Cecal Ligation & Puncture-induced plasma cytokine elevation and substantially reduce the severity of lung inflammation post intratracheal P. aeruginosa infection in mice (100μg/mouse; i.p.) in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Description 98% by HPLC
Target Proteins Fbox3
Molecular Formula C26H26N4
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.