Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Fetuin, Fetal Bovine Serum

Catalog No. 3415061GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-150-61GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-150-61GM Supplier MilliporeSigma™ Supplier No. 3415061GM
Add to Cart
Add to Cart

Promotes cell attachment, growth, and differentiation in many different culture systems

  • α-globulin that inhibits trypsin activity
  • Contains both N- and O-glycosidically-linked carbohydrates
  • Functions as opsonin for cationic-deactivating molecules in macrophages
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.5% (NH4)2SO4.
Quantity 1 g
Storage Requirements -20°C
Product Type Fetuin, Fetal Bovine Serum
Form Lyophilized
Purity or Quality Grade ≥70% by electrophoresis

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.