Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ FGF/VEGF Receptor Tyrosine Kinase Inhibitor, PD173074

Catalog No. 3416075MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-160-75MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-160-75MG Supplier MilliporeSigma™ Supplier No. 3416075MG
Add to Cart
Add to Cart

The FGF/VEGF Receptor Tyrosine Kinase Inhibitor, PD173074 controls the biological activity of FGF/VEGF Receptor Tyrosine Kinase. Primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by HPLC
CAS 219580-11-7
Molecular Formula C28H41N7O3
Formula Weight 523.7g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.