Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ FK-506, Streptomyces sp.

Catalog No. 3425005MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-250-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-250-05MG Supplier MilliporeSigma™ Supplier No. 3425005MG
Add to Cart
Add to Cart

Immunosuppressant that blocks T cell proliferation in vitro by inhibiting the generation of several lymphokines, especially IL-2.

Immunosuppressant that blocks T cell proliferation in vitro by inhibiting the generation of several lymphokines, especially IL-2. Shown to inhibit the activity of FK-506 binding protein, thereby reversing its effects on sarcoplasmic reticulum Ca2+ release. Inhibits the Ca2+-dependent phosphatase, Protein Phosphatase 2B (Calcineurin; Cat. No. 539568), as well as Na+,K+-ATPase in nephron segments. Also shown to inhibit aldosterone-induced synthesis of Giα-3 protein.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by TLC
Target Proteins Immunosuppressant that blocks T cell proliferation
CAS 104987-11-3
Formula Weight 804.1g/mol
Solubility DMSO or MeOH
Content And Storage Protect from moisture
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.